siRNA Details
| Gene Symbol: CD58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017002869.1 | XP_016858358.1 | 6 | 7055 | ||
| Gene Symbol: LOC100506459 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK126833.1 | - | 1 | 264 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000456414.2 | - | 1 | 264 | Chr. 1 - 116493023 - 116499212 [-] on Build GRCh38 | |

CD58
Genomic Map