siRNA Details
| Gene Symbol: LINC01266 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110118.1 | 11 | ||||
| GenBank mRNA | AK126307.1 | - | 1, 2 | 108 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000420823.5 | - | 1, 2 | 108 | Chr. 3 - 536231 - 846015 [+] on Build GRCh38 | |

LINC01266
Genomic Map