siRNA Details
| Gene Symbol: LOC101929395 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110611.1 | 2 | 417 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000417299.1 | - | 2 | 417 | Chr. 20 - 10753090 - 10753966 [+] on Build GRCh38 | |

LOC101929395
Genomic Map