siRNA Details
Gene Symbol: LINC01526 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126363.1 | 2 | 198 | |||
GenBank mRNA | BC038196.1 | - | 2 | 198 | ||
BC038576.1 | - | 2 | 198 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000441949.1 | - | 2 | 163 | Chr. 6 - 81813286 - 81814157 [-] on Build GRCh38 |
