siRNA Details
| Gene Symbol: LOC102723566 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136302.1 | 1 | 1098 | |||
| GenBank mRNA | AK057719.1 | - | 1 | 1098 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000439298.5 | - | 1 | 1098 | Chr. 9 - 127816066 - 127822520 [+] on Build GRCh38 | |

LOC102723566
Genomic Map