siRNA Details
| Gene Symbol: RREB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK022160.1 | - | 1 | 433 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000451355.2 | - | 1 | 433 | Chr. 6 - 7183083 - 7185287 [+] on Build GRCh38 | |

RREB1
Genomic Map