siRNA Details
| Gene Symbol: PLEKHM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006712521.3 | XP_006712584.1 | 7 | 2760 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000421964.1 | - | 2 | 428 | Chr. 2 - 207868582 - 207869915 [-] on Build GRCh38 | |

PLEKHM3
Genomic Map