siRNA Details
Gene Symbol: OSTM1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC031282.1 | - | 1 | 1331 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000441184.1 | - | 2 | 2953 | Chr. 6 - 108123633 - 108159392 [+] on Build GRCh38 |
