siRNA Details
| Gene Symbol: OSTM1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF520419.1 | - | 2 | 914 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000441184.1 | - | 2 | 917 | Chr. 6 - 108123633 - 108159392 [+] on Build GRCh38 | |

OSTM1-AS1
Genomic Map