siRNA Details
| Gene Symbol: LINC00457 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_047036.1 | 3 | 372 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000440160.1 | - | 3 | 372 | Chr. 13 - 34435450 - 34640685 [-] on Build GRCh38 | |

LINC00457
Genomic Map