siRNA Details
| Gene Symbol: TAF1A-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110613.1 | 1, 2 | 42 | |||
| GenBank mRNA | HY020214.1 | - | 1, 2 | 42 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TAF1A-AS1| Gene Symbol: TAF1A-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110613.1 | 1, 2 | 42 | |||
| GenBank mRNA | HY020214.1 | - | 1, 2 | 42 | ||