siRNA Details
| Gene Symbol: FREM2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | CK903491.1 | - | 1 | 102 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000448887.1 | - | 1 | 81 | Chr. 13 - 38821798 - 38827570 [-] on Build GRCh38 | |

FREM2-AS1
Genomic Map