siRNA Details
| Gene Symbol: TMEM246-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121573.1 | 3, 4 | 429 | |||
| NR_121574.1 | 2, 3 | 391 | ||||
| NR_121576.1 | 2, 3 | 262 | ||||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000425734.1 | - | 2, 3 | 334 | Chr. 9 - 101469379 - 101481502 [+] on Build GRCh38 | |

TMEM246-AS1
Genomic Map