siRNA Details
Gene Symbol: TMEM246-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121573.1 | 4 | 502 | |||
NR_121574.1 | 3 | 464 | ||||
NR_121575.1 | 2 | 380 | ||||
NR_121576.1 | 3 | 335 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000425734.1 | - | 3 | 407 | Chr. 9 - 101469379 - 101481502 [+] on Build GRCh38 |
