siRNA Details
Gene Symbol: TMEM246-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121574.1 | 1 | 69 | |||
NR_121575.1 | 1 | 69 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000425734.1 | - | 1 | 12 | Chr. 9 - 101469379 - 101481502 [+] on Build GRCh38 |
