siRNA Details
Gene Symbol: NRSN2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109990.1 | 3 | 655 | |||
GenBank mRNA | BF798129.1 | - | 1 | 135 | ||
BX104512.1 | - | 3 | 653 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000414676.5 | - | 3 | 637 | Chr. 20 - 320313 - 325234 [-] on Build GRCh38 |
