siRNA Details
Gene Symbol: NRSN2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109990.1 | 1 | 80 | |||
GenBank mRNA | BM849624.1 | - | 1 | 80 | ||
BX104512.1 | - | 1 | 78 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: NRSN2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109990.1 | 1 | 80 | |||
GenBank mRNA | BM849624.1 | - | 1 | 80 | ||
BX104512.1 | - | 1 | 78 |