siRNA Details
| Gene Symbol: LOC101928233 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_131192.1 | 2 | 525 | |||
| GenBank mRNA | BC041455.1 | - | 2 | 488 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000419628.1 | - | 2 | 488 | Chr. 21 - 42599280 - 42615058 [-] on Build GRCh38 | |

LOC101928233
Genomic Map