siRNA Details
| Gene Symbol: LINC01504 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110952.1 | 5 | 2013 | |||
| GenBank mRNA | AK095210.1 | - | 5 | 2013 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000436054.1 | - | 5 | 2013 | Chr. 9 - 72305436 - 72343210 [+] on Build GRCh38 | |

LINC01504
Genomic Map