siRNA Details
| Gene Symbol: PACRG-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110871.1 | 2 | 790 | |||
| NR_110872.1 | 2 | 790 | ||||
| GenBank mRNA | AK058177.1 | - | 2 | 790 | ||
| BC033504.1 | - | 2 | 775 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000418665.1 | - | 2 | 790 | Chr. 6 - 163042980 - 163054161 [-] on Build GRCh38 | |

PACRG-AS2
Genomic Map