siRNA Details
Gene Symbol: PACRG-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110871.1 | 2 | 399 | |||
NR_110872.1 | 2 | 399 | ||||
GenBank mRNA | AK058177.1 | - | 2 | 399 | ||
BC033504.1 | - | 2 | 386 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: PACRG-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110871.1 | 2 | 399 | |||
NR_110872.1 | 2 | 399 | ||||
GenBank mRNA | AK058177.1 | - | 2 | 399 | ||
BC033504.1 | - | 2 | 386 |