siRNA Details
| Gene Symbol: LOC105374596 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135239.1 | 3 | 132 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000435357.1 | - | 2 | 114 | Chr. 2 - 52373118 - 52390012 [+] on Build GRCh38 | |

LOC105374596
Genomic Map