siRNA Details
| Gene Symbol: LOC100240728 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108095.1 | 1 | 155 | |||
| GenBank mRNA | BC036258.1 | - | 1 | 131 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000436042.1 | - | 1 | 131 | Chr. 7 - 56482105 - 56483295 [-] on Build GRCh38 | |

LOC100240728
Genomic Map