siRNA Details
Gene Symbol: LINC00475 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027341.1 | 4 | 1946 | |||
GenBank mRNA | AK023662.1 | - | 4 | 1946 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000416438.6 | - | 4 | 1946 | Chr. 9 - 92141467 - 92160114 [+] on Build GRCh38 |
