siRNA Details
| Gene Symbol: LINC01393 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120521.1 | 4 | 977 | |||
| GenBank mRNA | BC022431.1 | - | 4 | 976 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000435981.5 | - | 4 | 977 | Chr. 7 - 115078958 - 115126314 [+] on Build GRCh38 | |

LINC01393
Genomic Map