siRNA Details
| Gene Symbol: LVCAT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_131961.1 | 4 | 1544 | |||
| GenBank mRNA | CB751600.1 | - | 4 | 1523 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000426413.1 | - | 4 | 1544 | Chr. 7 - 119750815 - 119907375 [-] on Build GRCh38 | |

LVCAT5
Genomic Map