siRNA Details
Gene Symbol: LVCAT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_131961.1 | 4 | 1510 | |||
GenBank mRNA | CB751600.1 | - | 4 | 1489 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000426413.1 | - | 4 | 1510 | Chr. 7 - 119750815 - 119907375 [-] on Build GRCh38 |
