siRNA Details
Gene Symbol: LINC01492 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121578.1 | 10 | 2744 | |||
GenBank mRNA | BC035187.1 | - | 10 | 2751 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000411575.5 | - | 10 | 2745 | Chr. 9 - 103140523 - 103325034 [-] on Build GRCh38 |
