siRNA Details
| Gene Symbol: DOCK4-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103806.1 | 2 | 1165 | |||
| GenBank mRNA | BC043243.2 | - | 2 | 1134 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000452714.1 | - | 2 | 1154 | Chr. 7 - 111808516 - 111821773 [+] on Build GRCh38 | |

DOCK4-AS1
Genomic Map