siRNA Details
| Gene Symbol: LOC729083 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_122070.1 | 1 | 190 | |||
| GenBank mRNA | BC038366.1 | - | 1 | 190 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000432377.1 | - | 1 | 190 | Chr. 3 - 42770612 - 42773635 [+] on Build GRCh38 | |

LOC729083
Genomic Map