siRNA Details
Gene Symbol: LINCMD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_131248.1 | 2 | 167 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINCMD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_131248.1 | 2 | 167 |