siRNA Details
| Gene Symbol: C9orf135-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038833.1 | 2 | 267 | |||
| GenBank mRNA | BC039385.1 | - | 2 | 289 | ||
| BG717523.1 | - | 2 | 245 | |||
| BI830190.1 | - | 2 | 267 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C9orf135-AS1| Gene Symbol: C9orf135-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038833.1 | 2 | 267 | |||
| GenBank mRNA | BC039385.1 | - | 2 | 289 | ||
| BG717523.1 | - | 2 | 245 | |||
| BI830190.1 | - | 2 | 267 | |||