siRNA Details
| Gene Symbol: PTPRD-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121599.1 | 3 | 1186 | |||
| NR_121600.1 | 2 | 1104 | ||||
| GenBank mRNA | AK094342.1 | - | 2 | 1814 | ||
| DB541705.1 | - | 1 | 310 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PTPRD-AS1| Gene Symbol: PTPRD-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121599.1 | 3 | 1186 | |||
| NR_121600.1 | 2 | 1104 | ||||
| GenBank mRNA | AK094342.1 | - | 2 | 1814 | ||
| DB541705.1 | - | 1 | 310 | |||