siRNA Details
| Gene Symbol: NRG3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120666.1 | 1 | 102 | |||
| GenBank mRNA | AA383013.1 | - | 1 | 37 | ||
| CR737767.1 | - | 1 | 37 | |||
| DB052745.1 | - | 1 | 101 | |||
| DB082602.1 | - | 1 | 103 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000448936.2 | - | 1 | 101 | Chr. 10 - 82230130 - 82232919 [-] on Build GRCh38 | |

NRG3-AS1
Genomic Map