siRNA Details
| Gene Symbol: DBH-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102735.1 | 2 | 821 | |||
| GenBank mRNA | AF129263.1 | - | 1 | 1493 | ||
| AF129264.1 | - | 2 | 726 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000425189.1 | - | 2 | 726 | Chr. 9 - 133654586 - 133657313 [-] on Build GRCh38 | |

DBH-AS1
Genomic Map