siRNA Details
| Gene Symbol: LOC105375304 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136270.1 | 2 | 525 | |||
| NR_136271.1 | 2 | 437 | ||||
| NR_136272.1 | 2 | 332 | ||||
| GenBank mRNA | BG208557.1 | - | 1 | 261 | ||
| CK824920.1 | - | 2 | 390 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105375304| Gene Symbol: LOC105375304 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136270.1 | 2 | 525 | |||
| NR_136271.1 | 2 | 437 | ||||
| NR_136272.1 | 2 | 332 | ||||
| GenBank mRNA | BG208557.1 | - | 1 | 261 | ||
| CK824920.1 | - | 2 | 390 | |||