siRNA Details
| Gene Symbol: LINC00626 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024160.1 | 3 | 804 | |||
| GenBank mRNA | BC068047.1 | - | 3 | 671 | ||
| BC082964.1 | - | 3 | 671 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000420691.1 | - | 3 | 806 | Chr. 1 - 168786939 - 168792886 [+] on Build GRCh38 | |

LINC00626
Genomic Map