siRNA Details
| Gene Symbol: LOC102724231 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134942.1 | 2 | 528 | |||
| NR_134943.1 | 2 | 520 | ||||
| GenBank mRNA | BX377745.2 | - | 2 | 502 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102724231| Gene Symbol: LOC102724231 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134942.1 | 2 | 528 | |||
| NR_134943.1 | 2 | 520 | ||||
| GenBank mRNA | BX377745.2 | - | 2 | 502 | ||