siRNA Details
| Gene Symbol: LOC102724231 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134942.1 | 2 | 399 | |||
| NR_134943.1 | 2 | 391 | ||||
| GenBank mRNA | AA448155.1 | - | 2 | 437 | ||
| BX377745.2 | - | 2 | 373 | |||
| HY025704.1 | - | 2 | 392 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102724231| Gene Symbol: LOC102724231 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134942.1 | 2 | 399 | |||
| NR_134943.1 | 2 | 391 | ||||
| GenBank mRNA | AA448155.1 | - | 2 | 437 | ||
| BX377745.2 | - | 2 | 373 | |||
| HY025704.1 | - | 2 | 392 | |||