siRNA Details
| Gene Symbol: LOC100506937 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134332.1 | 1 | 315 | |||
| GenBank mRNA | BX103078.1 | - | 1 | 315 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000436379.1 | - | 1 | 315 | Chr. 7 - 132758970 - 132760632 [+] on Build GRCh38 | |

LOC100506937
Genomic Map