siRNA Details
Gene Symbol: LOC101928324 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125952.1 | 3 | 633 | |||
NR_125953.1 | 2 | 512 | ||||
GenBank mRNA | AL050085.1 | - | 2 | 512 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000423060.1 | - | 3 | 378 | Chr. 1 - 26462756 - 26467282 [-] on Build GRCh38 |
