siRNA Details
| Gene Symbol: LOC101929163 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK097727.1 | - | 1 | 670 | ||
| AK123889.1 | - | 2 | 1472 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000425033.1 | - | 2 | 1471 | Chr. 6 - 32255711 - 32265838 [+] on Build GRCh38 | |

LOC101929163
Genomic Map