siRNA Details
| Gene Symbol: PCA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015342.2 | 3 | 1333 | |||
| NR_132312.1 | 4 | 1498 | ||||
| NR_132313.1 | 3 | 1198 | ||||
| GenBank mRNA | AY894120.1 | - | 3 | 1331 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000412654.1 | - | 4 | 1498 | Chr. 9 - 76764436 - 76787569 [+] on Build GRCh38 | |

PCA3
Genomic Map