siRNA Details
| Gene Symbol: LINC01534 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110707.1 | 3 | 700 | |||
| GenBank mRNA | BQ049787.1 | - | 3 | 661 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000425254.2 | - | 3 | 661 | Chr. 19 - 36685465 - 36687410 [-] on Build GRCh38 | |

LINC01534
Genomic Map