siRNA Details
| Gene Symbol: LAMP5-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109957.1 | 5 | 1326 | |||
| GenBank mRNA | AK096264.1 | - | 5 | 1326 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000443469.1 | - | 5 | 1326 | Chr. 20 - 9505180 - 9514998 [-] on Build GRCh38 | |

LAMP5-AS1
Genomic Map