siRNA Details
| Gene Symbol: ZNF503-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038223.1 | 3 | 1430 | |||
| NR_038224.1 | 3 | 1088 | ||||
| NR_038225.1 | 4 | 1199 | ||||
| NR_038226.1 | 4 | 1211 | ||||
| GenBank mRNA | AK023445.1 | - | 3 | 1423 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000416398.1 | - | 3 | 1423 | Chr. 10 - 75296390 - 75361678 [+] on Build GRCh38 | |

ZNF503-AS1
Genomic Map