siRNA Details
| Gene Symbol: LOC101927769 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110117.1 | 5 | 803 | |||
| GenBank mRNA | AK026386.1 | - | 2 | 673 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000455373.1 | - | 2 | 670 | Chr. 7 - 20305711 - 20311712 [+] on Build GRCh38 | |

LOC101927769
Genomic Map