siRNA Details
| Gene Symbol: AP4B1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037864.1 | 2, 3 | 223 | |||
| GenBank mRNA | AK123199.1 | - | 2, 3 | 222 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AP4B1-AS1| Gene Symbol: AP4B1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037864.1 | 2, 3 | 223 | |||
| GenBank mRNA | AK123199.1 | - | 2, 3 | 222 | ||