siRNA Details
| Gene Symbol: LEMD1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038425.1 | 4 | 1981 | |||
| GenBank mRNA | AK090497.1 | - | 4 | 1982 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000447832.1 | - | 4 | 1981 | Chr. 1 - 205373252 - 205387440 [+] on Build GRCh38 | |

LEMD1-AS1
Genomic Map