siRNA Details
| Gene Symbol: LOC101929541 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125983.1 | 3 | 487 | |||
| NR_125984.1 | 4 | 366 | ||||
| GenBank mRNA | BX099738.1 | - | 4 | 366 | ||
| F26527.1 | - | 3 | 297 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000412221.5 | - | 2 | 299 | Chr. 1 - 212225280 - 212226538 [-] on Build GRCh38 | |

LOC101929541
Genomic Map